Product Details

SNP ID
rs78772596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49842583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCAGAACTCATTTTGTAGAAGA[A/G]GAAACTAAGGCCAAAAGTGCTTGGT
Phenotype
MIM: 607833
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ45513 PubMed Links

Gene Details

Gene
FLJ45513
Gene Name
uncharacterized LOC729220
There are no transcripts associated with this gene.

Gene
TAC4
Gene Name
tachykinin 4 (hemokinin)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077503.1 Intron NP_001070971.1
NM_001077504.1 Intron NP_001070972.1
NM_001077505.1 Intron NP_001070973.1
NM_001077506.1 Intron NP_001070974.1
NM_170685.2 Intron NP_733786.2

View Full Product Details