Product Details
- SNP ID
-
rs79885699
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78234612 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCAGTGCTGCACCGTGCAGTCAT[G/T]GTGGTCCTGCCCCTGAGCCTGGTCC
- Phenotype
-
MIM: 603352
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11077350] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BIRC5
- Gene Name
- baculoviral IAP repeat containing 5
There are no transcripts associated with this gene.
- Gene
- TMEM235
- Gene Name
- transmembrane protein 235
View Full Product Details