Product Details

SNP ID
rs78633734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14074264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTGACGCATGTTCAGCCAAAG[A/G]AGCAGACAGATCAAGAAATTGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF519 PubMed Links

Gene Details

Gene
ZNF519
Gene Name
zinc finger protein 519
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145287.3 13440 Intron NP_660330.2
XM_011525605.2 13440 Intron XP_011523907.1
XM_017025562.1 13440 UTR 3 XP_016881051.1
XM_017025563.1 13440 UTR 3 XP_016881052.1
XM_017025564.1 13440 UTR 3 XP_016881053.1
XM_017025565.1 13440 UTR 3 XP_016881054.1
XM_017025566.1 13440 Intron XP_016881055.1

View Full Product Details