Product Details

SNP ID
rs80008114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5393025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGAAAATCTGACACACCGGAC[A/G]GACCTAGACAGCTTCTAGCATTTGA
Phenotype
MIM: 605331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41L3 PubMed Links

Gene Details

Gene
EPB41L3
Gene Name
erythrocyte membrane protein band 4.1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281533.1 3404 UTR 3 NP_001268462.1
NM_001281534.1 3404 UTR 3 NP_001268463.1
NM_001281535.1 3404 UTR 3 NP_001268464.1
NM_012307.3 3404 UTR 3 NP_036439.2
XM_011525623.2 3404 UTR 3 XP_011523925.1
XM_011525626.2 3404 UTR 3 XP_011523928.1
XM_011525628.2 3404 UTR 3 XP_011523930.1
XM_011525632.2 3404 UTR 3 XP_011523934.1
XM_011525634.2 3404 UTR 3 XP_011523936.1
XM_011525637.2 3404 UTR 3 XP_011523939.1
XM_017025618.1 3404 UTR 3 XP_016881107.1
XM_017025619.1 3404 UTR 3 XP_016881108.1
XM_017025620.1 3404 UTR 3 XP_016881109.1
XM_017025621.1 3404 UTR 3 XP_016881110.1
XM_017025622.1 3404 UTR 3 XP_016881111.1
XM_017025623.1 3404 UTR 3 XP_016881112.1
XM_017025624.1 3404 UTR 3 XP_016881113.1
XM_017025625.1 3404 UTR 3 XP_016881114.1
XM_017025626.1 3404 UTR 3 XP_016881115.1
XM_017025627.1 3404 UTR 3 XP_016881116.1
XM_017025628.1 3404 UTR 3 XP_016881117.1
XM_017025629.1 3404 UTR 3 XP_016881118.1
XM_017025630.1 3404 UTR 3 XP_016881119.1
XM_017025631.1 3404 UTR 3 XP_016881120.1
XM_017025632.1 3404 UTR 3 XP_016881121.1
XM_017025633.1 3404 UTR 3 XP_016881122.1
XM_017025634.1 3404 UTR 3 XP_016881123.1
XM_017025635.1 3404 UTR 3 XP_016881124.1
XM_017025636.1 3404 UTR 3 XP_016881125.1
XM_017025637.1 3404 UTR 3 XP_016881126.1
XM_017025638.1 3404 UTR 3 XP_016881127.1
XM_017025639.1 3404 UTR 3 XP_016881128.1
XM_017025640.1 3404 UTR 3 XP_016881129.1
XM_017025641.1 3404 UTR 3 XP_016881130.1
XM_017025642.1 3404 UTR 3 XP_016881131.1
XM_017025643.1 3404 UTR 3 XP_016881132.1
XM_017025644.1 3404 UTR 3 XP_016881133.1
XM_017025645.1 3404 UTR 3 XP_016881134.1
XM_017025646.1 3404 UTR 3 XP_016881135.1
XM_017025647.1 3404 UTR 3 XP_016881136.1
XM_017025648.1 3404 UTR 3 XP_016881137.1
XM_017025649.1 3404 UTR 3 XP_016881138.1
XM_017025650.1 3404 UTR 3 XP_016881139.1
XM_017025651.1 3404 UTR 3 XP_016881140.1
XM_017025652.1 3404 UTR 3 XP_016881141.1
XM_017025653.1 3404 UTR 3 XP_016881142.1
XM_017025654.1 3404 UTR 3 XP_016881143.1
XM_017025655.1 3404 UTR 3 XP_016881144.1
XM_017025656.1 3404 UTR 3 XP_016881145.1
XM_017025657.1 3404 UTR 3 XP_016881146.1
XM_017025658.1 3404 UTR 3 XP_016881147.1
XM_017025659.1 3404 UTR 3 XP_016881148.1
XM_017025660.1 3404 UTR 3 XP_016881149.1

View Full Product Details