Product Details

SNP ID
rs76152118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40294809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAGTGACAAGCGGCATTCTCT[C/T]TGTCCACTCGTGCCAGCTCTCAGGA
Phenotype
MIM: 164731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKT2 PubMed Links

Gene Details

Gene
AKT2
Gene Name
AKT serine/threonine kinase 2
There are no transcripts associated with this gene.

Gene
C19orf47
Gene Name
chromosome 19 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256440.1 1657 Intron NP_001243369.1
NM_001256441.1 1657 Intron NP_001243370.1
XM_005258520.2 1657 Intron XP_005258577.1
XM_011526460.2 1657 UTR 3 XP_011524762.1
XM_017026291.1 1657 Intron XP_016881780.1
XM_017026292.1 1657 Intron XP_016881781.1
XM_017026293.1 1657 Intron XP_016881782.1
XM_017026294.1 1657 Intron XP_016881783.1
XM_017026295.1 1657 Intron XP_016881784.1

View Full Product Details