Product Details

SNP ID
rs79507858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51078833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTACCTGACAAGAGTCCTTCCC[A/G]CCCTGGGGAACTCCTGCACAGACCA
Phenotype
MIM: 606135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK14 PubMed Links

Gene Details

Gene
KLK14
Gene Name
kallikrein related peptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311182.1 817 Silent Mutation NP_001298111.1
NM_022046.5 817 Silent Mutation NP_071329.2
XM_017026820.1 817 UTR 3 XP_016882309.1

View Full Product Details