Product Details

SNP ID
rs79139857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36512302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAATGTAAGTCCATTATGTTTC[A/C]TGTACAGCTTCCTCACTCCTATCTG
Phenotype
MIM: 613749
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012756.2 3853 UTR 3 NP_001012774.1
NM_001166036.1 3853 UTR 3 NP_001159508.1
NM_001166037.1 3853 UTR 3 NP_001159509.1
NM_001166038.1 3853 UTR 3 NP_001159510.1
XM_011526897.2 3853 UTR 3 XP_011525199.1
XM_017026741.1 3853 UTR 3 XP_016882230.1

View Full Product Details