Product Details

SNP ID
rs74317014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4174862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCACACGGGGGCCGTCCCAGG[C/T]GGGGATCTCCAGCCCCAGGTGCTTC
Phenotype
MIM: 611998 MIM: 606211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD24 PubMed Links

Gene Details

Gene
ANKRD24
Gene Name
ankyrin repeat domain 24
There are no transcripts associated with this gene.

Gene
CREB3L3
Gene Name
cAMP responsive element binding protein 3 like 3
There are no transcripts associated with this gene.

Gene
SIRT6
Gene Name
sirtuin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193285.2 705 Missense Mutation ACC,GCC T248A NP_001180214.1
NM_001321058.1 705 Missense Mutation ACC,GCC T203A NP_001307987.1
NM_001321059.1 705 Missense Mutation ACC,GCC T214A NP_001307988.1
NM_001321060.1 705 UTR 3 NP_001307989.1
NM_001321061.1 705 Missense Mutation ACC,GCC T176A NP_001307990.1
NM_001321062.1 705 Missense Mutation ACC,GCC T140A NP_001307991.1
NM_001321063.1 705 UTR 3 NP_001307992.1
NM_001321064.1 705 UTR 3 NP_001307993.1
NM_016539.3 705 Missense Mutation ACC,GCC T275A NP_057623.2

View Full Product Details