Product Details

SNP ID
rs75098773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14088605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGACAGGAACAGAGAGAGCCGG[A/G]ATCAGGGCACATCCCAGGGAGGCCC
Phenotype
MIM: 601639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf67 PubMed Links

Gene Details

Gene
C19orf67
Gene Name
chromosome 19 open reading frame 67
There are no transcripts associated with this gene.

Gene
PRKACA
Gene Name
protein kinase cAMP-activated catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
SAMD1
Gene Name
sterile alpha motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138352.1 Intron NP_612361.1

View Full Product Details