Product Details

SNP ID
rs80352265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18293308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTGGAACATGCAAGATGGTTCC[A/G]TCACACATTTGACATATTTTCGTGG
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 3491 Intron NP_001076799.1
NM_001282995.1 3491 Intron NP_001269924.1
NM_001282996.1 3491 Intron NP_001269925.1
NM_001282997.1 3491 Intron NP_001269926.1
NM_001282998.1 3491 Intron NP_001269927.1
NM_001282999.1 3491 Intron NP_001269928.1
NM_001283000.1 3491 Intron NP_001269929.1
NM_001283001.1 3491 Intron NP_001269930.1
NM_001283002.1 3491 Intron NP_001269931.1
NM_001283003.1 3491 Intron NP_001269932.1
NM_001283004.1 3491 Intron NP_001269933.1
NM_001283005.1 3491 Intron NP_001269934.1
NM_001283006.1 3491 Intron NP_001269935.1
NM_001283007.1 3491 Intron NP_001269936.1
NM_001283008.1 3491 Intron NP_001269937.1
NM_003434.5 3491 Intron NP_003425.2
XM_005260819.1 3491 Intron XP_005260876.1
XM_005260820.4 3491 Intron XP_005260877.1
XM_011529336.1 3491 Intron XP_011527638.1
XM_011529337.1 3491 Intron XP_011527639.1
XM_011529338.2 3491 Intron XP_011527640.1
XM_011529339.1 3491 Intron XP_011527641.1
XM_011529340.1 3491 Intron XP_011527642.1
XM_017028041.1 3491 UTR 5 XP_016883530.1
XM_017028042.1 3491 Intron XP_016883531.1
XM_017028043.1 3491 UTR 5 XP_016883532.1
XM_017028044.1 3491 UTR 5 XP_016883533.1
XM_017028045.1 3491 UTR 5 XP_016883534.1
XM_017028046.1 3491 UTR 5 XP_016883535.1
XM_017028047.1 3491 Intron XP_016883536.1
XM_017028048.1 3491 Intron XP_016883537.1
XM_017028049.1 3491 UTR 5 XP_016883538.1
XM_017028050.1 3491 Intron XP_016883539.1
XM_017028051.1 3491 Intron XP_016883540.1
XM_017028052.1 3491 Intron XP_016883541.1
XM_017028053.1 3491 Intron XP_016883542.1
XM_017028054.1 3491 Intron XP_016883543.1
XM_017028055.1 3491 Intron XP_016883544.1
XM_017028056.1 3491 Intron XP_016883545.1
XM_017028057.1 3491 Intron XP_016883546.1
XM_017028058.1 3491 Intron XP_016883547.1

View Full Product Details