Product Details

SNP ID
rs76116142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18298045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGGAAAAAAAGCCACAGGGTCC[C/T]GGAGAGCCAGGGGAATGGTGAGTGT
Phenotype
MIM: 604075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 116 Intron NP_001076799.1
NM_001282995.1 116 UTR 5 NP_001269924.1
NM_001282996.1 116 UTR 5 NP_001269925.1
NM_001282997.1 116 Intron NP_001269926.1
NM_001282998.1 116 Intron NP_001269927.1
NM_001282999.1 116 Intron NP_001269928.1
NM_001283000.1 116 Intron NP_001269929.1
NM_001283001.1 116 Intron NP_001269930.1
NM_001283002.1 116 UTR 5 NP_001269931.1
NM_001283003.1 116 Intron NP_001269932.1
NM_001283004.1 116 Intron NP_001269933.1
NM_001283005.1 116 Intron NP_001269934.1
NM_001283006.1 116 Intron NP_001269935.1
NM_001283007.1 116 Intron NP_001269936.1
NM_001283008.1 116 Intron NP_001269937.1
NM_003434.5 116 UTR 5 NP_003425.2
XM_005260819.1 116 Intron XP_005260876.1
XM_005260820.4 116 Intron XP_005260877.1
XM_011529336.1 116 UTR 5 XP_011527638.1
XM_011529337.1 116 UTR 5 XP_011527639.1
XM_011529338.2 116 UTR 5 XP_011527640.1
XM_011529339.1 116 UTR 5 XP_011527641.1
XM_011529340.1 116 Intron XP_011527642.1
XM_017028041.1 116 UTR 5 XP_016883530.1
XM_017028042.1 116 UTR 5 XP_016883531.1
XM_017028043.1 116 UTR 5 XP_016883532.1
XM_017028044.1 116 UTR 5 XP_016883533.1
XM_017028045.1 116 UTR 5 XP_016883534.1
XM_017028046.1 116 UTR 5 XP_016883535.1
XM_017028047.1 116 Intron XP_016883536.1
XM_017028048.1 116 Intron XP_016883537.1
XM_017028049.1 116 UTR 5 XP_016883538.1
XM_017028050.1 116 UTR 5 XP_016883539.1
XM_017028051.1 116 UTR 5 XP_016883540.1
XM_017028052.1 116 UTR 5 XP_016883541.1
XM_017028053.1 116 Intron XP_016883542.1
XM_017028054.1 116 Intron XP_016883543.1
XM_017028055.1 116 UTR 5 XP_016883544.1
XM_017028056.1 116 UTR 5 XP_016883545.1
XM_017028057.1 116 UTR 5 XP_016883546.1
XM_017028058.1 116 UTR 5 XP_016883547.1

View Full Product Details