Product Details

SNP ID
rs75023887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143884255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATTCCCCCTTTGAGGGAGGGA[C/G]TTTTAAACGTGAACTATTACTTGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UBE2NL PubMed Links

Gene Details

Gene
UBE2NL
Gene Name
ubiquitin conjugating enzyme E2 N like (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012989.2 Intron NP_001013007.1

View Full Product Details