Product Details

SNP ID
rs80022851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:124666269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACCTTCCTTGTCATGAATTTC[C/T]CTCTTAACCCTGGCATCCAGCCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD50 PubMed Links

Gene Details

Gene
ANKRD50
Gene Name
ankyrin repeat domain 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167882.1 7310 UTR 3 NP_001161354.1
NM_020337.2 7310 UTR 3 NP_065070.1
XM_017008471.1 7310 UTR 3 XP_016863960.1

View Full Product Details