Product Details
- SNP ID
-
rs74523017
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:124668314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATTTATCATTTTTCACGTCTATG[C/T]CTTATGTGTGAAAACATTACTTAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD50
PubMed Links
Gene Details
- Gene
- ANKRD50
- Gene Name
- ankyrin repeat domain 50
View Full Product Details