Product Details

SNP ID
rs75717674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152626133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAATGGGTATTTTTACATATTT[A/C]TTAAAACAAGTTTCCTGAGAAGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM154 PubMed Links

Gene Details

Gene
TMEM154
Gene Name
transmembrane protein 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152680.2 3197 UTR 3 NP_689893.1
XM_011531716.2 3197 Intron XP_011530018.1

View Full Product Details