Product Details

SNP ID
rs74997268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127301131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGACAAGTCCTGTACATATAG[C/T]AGAAGCAGATACTTCCTTATGTTCC
Phenotype
MIM: 133510 MIM: 609487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERCC3 PubMed Links

Gene Details

Gene
ERCC3
Gene Name
ERCC excision repair 3, TFIIH core complex helicase subunit
There are no transcripts associated with this gene.

Gene
MAP3K2
Gene Name
mitogen-activated protein kinase kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006609.4 8408 UTR 3 NP_006600.3

View Full Product Details