Product Details
- SNP ID
-
rs78322232
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127701539 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGCCTTGTCCAAGATGGCGGACC[C/T]CCACCGCCAGCTGCAGGAGTACCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFT2D3
PubMed Links
Gene Details
- Gene
- SFT2D3
- Gene Name
- SFT2 domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032740.3 |
517 |
Missense Mutation |
CCC,CTC |
P4L |
NP_116129.3 |
- Gene
- WDR33
- Gene Name
- WD repeat domain 33
There are no transcripts associated with this gene.
View Full Product Details