Product Details

SNP ID
rs76605545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102631299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTAAAAGCATCTTTTCTTTT[C/T]TTTTTGTGCTAGTATTTATTTATTT
Phenotype
MIM: 600530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A2 PubMed Links

Gene Details

Gene
SLC9A2
Gene Name
solute carrier family 9 member A2
There are no transcripts associated with this gene.

View Full Product Details