Product Details

SNP ID
rs75573663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37345976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTAATGATGTCAGCATAAAATAG[C/T]ATGTTATAGAGATATTAAGAAAAGC
Phenotype
MIM: 607065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
QPCT PubMed Links

Gene Details

Gene
QPCT
Gene Name
glutaminyl-peptide cyclotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012413.3 Intron NP_036545.1

View Full Product Details