Product Details

SNP ID
rs79229886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63126440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACCATTCTCATTTTACAGATGA[A/C]GAAACAGGTATAGAGGTGTGTAGCA
Phenotype
MIM: 613580
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DBIL5P2 PubMed Links

Gene Details

Gene
DBIL5P2
Gene Name
diazepam binding inhibitor-like 5 pseudogene 2
There are no transcripts associated with this gene.

Gene
WDPCP
Gene Name
WD repeat containing planar cell polarity effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042692.2 Intron NP_001036157.1
NM_015910.5 Intron NP_056994.3
XM_005264348.3 Intron XP_005264405.1
XM_011532881.2 Intron XP_011531183.1
XM_011532884.2 Intron XP_011531186.1
XM_011532886.2 Intron XP_011531188.1
XM_011532887.2 Intron XP_011531189.1
XM_011532890.2 Intron XP_011531192.1
XM_011532891.2 Intron XP_011531193.1
XM_017004251.1 Intron XP_016859740.1
XM_017004252.1 Intron XP_016859741.1
XM_017004253.1 Intron XP_016859742.1
XM_017004254.1 Intron XP_016859743.1

View Full Product Details