Product Details

SNP ID
rs74768407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:84981076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTACTAGAAAAAAAAAAACTGA[A/G]ATCAGAAGAGTGGGTCAGTTGTATT
Phenotype
MIM: 614719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCMF1 PubMed Links

Gene Details

Gene
KCMF1
Gene Name
potassium channel modulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020122.4 Intron NP_064507.3
XM_006712052.3 Intron XP_006712115.1
XM_011532990.1 Intron XP_011531292.1
XM_017004511.1 Intron XP_016860000.1

View Full Product Details