Product Details

SNP ID
rs76044409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37454657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTCAGGCACCAACTTAACTCT[A/G]CTGAAATCTTTTTAAAATAATGCTT
Phenotype
MIM: 603963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA9 PubMed Links

Gene Details

Gene
ITGA9
Gene Name
integrin subunit alpha 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002207.2 Intron NP_002198.2

View Full Product Details