Product Details

SNP ID
rs76959863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52248166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAATCCAGGGCCATCTTGGTG[C/T]GGAGAGATGAGATACGGCCACTGAG
Phenotype
MIM: 608979 MIM: 607433 MIM: 611059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929054 PubMed Links

Gene Details

Gene
LOC101929054
Gene Name
uncharacterized LOC101929054
There are no transcripts associated with this gene.

Gene
PPM1M
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122870.2 645 Missense Mutation CGG,TGG R30W NP_001116342.1
NM_144641.3 645 Missense Mutation CGG,TGG R242W NP_653242.3
XM_005264879.2 645 Missense Mutation CGG,TGG R242W XP_005264936.1
XM_005264880.3 645 Missense Mutation CGG,TGG R157W XP_005264937.1
XM_005264881.1 645 Missense Mutation CGG,TGG R81W XP_005264938.1
Gene
TWF2
Gene Name
twinfilin actin binding protein 2
There are no transcripts associated with this gene.

Gene
WDR82
Gene Name
WD repeat domain 82
There are no transcripts associated with this gene.

View Full Product Details