Product Details

SNP ID
rs75498935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27924392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGCGCCGGGTGGAGCTGGCTGT[C/T]ATGTTGAACTTGACCGAGAGACACA
Phenotype
MIM: 600733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDX1 PubMed Links

Gene Details

Gene
PDX1
Gene Name
pancreatic and duodenal homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000209.3 651 Silent Mutation GTC,GTT V181V NP_000200.1
Gene
PDX1-AS1
Gene Name
PDX1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details