Product Details

SNP ID
rs77770447
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:68674421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAAAGCAAGGATCTAGAATTCC[A/G]AATTTTAGAAGTTATAAAGAGAACT
Phenotype
MIM: 616102
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMX3 PubMed Links

Gene Details

Gene
TMX3
Gene Name
thioredoxin related transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019022.3 3875 UTR 3 NP_061895.3
XM_005266714.2 3875 UTR 3 XP_005266771.1
XM_011526033.2 3875 UTR 3 XP_011524335.1
XM_011526034.2 3875 Intron XP_011524336.1
XM_011526035.2 3875 Intron XP_011524337.1

View Full Product Details