Product Details

SNP ID
rs75555038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110102509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATGCTTACTTTAATAAATAAA[A/G]TGCTCAATTCTATACTGACAGGAGT
Phenotype
MIM: 605035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WASF1 PubMed Links

Gene Details

Gene
WASF1
Gene Name
WAS protein family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024934.1 Intron NP_001020105.1
NM_001024935.1 Intron NP_001020106.1
NM_001024936.1 Intron NP_001020107.1
NM_003931.2 Intron NP_003922.1
XM_005267203.3 Intron XP_005267260.1
XM_005267205.2 Intron XP_005267262.1
XM_005267206.2 Intron XP_005267263.1
XM_005267207.2 Intron XP_005267264.1
XM_011536233.1 Intron XP_011534535.1
XM_011536234.1 Intron XP_011534536.1
XM_011536235.1 Intron XP_011534537.1
XM_017011437.1 Intron XP_016866926.1
XM_017011438.1 Intron XP_016866927.1
XM_017011439.1 Intron XP_016866928.1
XM_017011440.1 Intron XP_016866929.1
XM_017011441.1 Intron XP_016866930.1
XM_017011442.1 Intron XP_016866931.1
XM_017011443.1 Intron XP_016866932.1
XM_017011444.1 Intron XP_016866933.1

View Full Product Details