Product Details

SNP ID
rs76451980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:157987783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATTATTATTAACTATAGTTACC[A/G]TGCTATGCAAGTTCGAAATTTACTT
Phenotype
MIM: 609410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNJ2 PubMed Links

Gene Details

Gene
SYNJ2
Gene Name
synaptojanin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178088.1 Intron NP_001171559.1
NM_003898.3 Intron NP_003889.1
XM_005267198.4 Intron XP_005267255.1
XM_005267199.2 Intron XP_005267256.1
XM_005267200.1 Intron XP_005267257.1
XM_006715592.3 Intron XP_006715655.1
XM_011536224.1 Intron XP_011534526.1
XM_011536225.1 Intron XP_011534527.1
XM_011536226.2 Intron XP_011534528.1
XM_011536227.2 Intron XP_011534529.1
XM_011536228.2 Intron XP_011534530.1
XM_011536230.2 Intron XP_011534532.1
XM_017011406.1 Intron XP_016866895.1

View Full Product Details