Product Details

SNP ID
rs78779496
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:168293869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTGACTGGTCAGTCAATGCA[A/G]CCCCCAGCTTGGTGTAGTGCTCTGT
Phenotype
MIM: 608966
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DACT2 PubMed Links
Additional Information
For this assay, SNP(s) [rs33979593] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DACT2
Gene Name
dishevelled binding antagonist of beta catenin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286350.1 989 Intron NP_001273279.1
NM_001286351.1 989 UTR 3 NP_001273280.1
NM_214462.4 989 Intron NP_999627.2
XM_011535507.2 989 Intron XP_011533809.1

View Full Product Details