Product Details

SNP ID
rs79422589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22947340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGACGGTCAAGGTGTGGGCTAA[A/G]CACTGATAGGATAAACAAGGTCACT
Phenotype
MIM: 613431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS4 PubMed Links

Gene Details

Gene
HAUS4
Gene Name
HAUS augmin like complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166269.1 Intron NP_001159741.1
NM_001166270.1 Intron NP_001159742.1
NM_017815.2 Intron NP_060285.2
Gene
LOC101926933
Gene Name
uncharacterized LOC101926933
There are no transcripts associated with this gene.

Gene
MIR4707
Gene Name
microRNA 4707
There are no transcripts associated with this gene.

View Full Product Details