Product Details

SNP ID
rs78875724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73478735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCCCACAGCAATCCTTTTCTC[G/T]GGGGAGGTGCGTAGATCTTTGACAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HEATR4 PubMed Links

Gene Details

Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 3782 Silent Mutation NP_001207413.1
NM_203309.2 3782 Silent Mutation NP_976054.2
XM_006720143.2 3782 Silent Mutation XP_006720206.1
XM_011536760.2 3782 Silent Mutation XP_011535062.1
XM_011536761.2 3782 Intron XP_011535063.1
XM_017021289.1 3782 Silent Mutation XP_016876778.1

View Full Product Details