Product Details

SNP ID
rs77284446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24080118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAAGCAGGGATAAAGAATGAA[C/T]ACTTCTTAAATTGCAAGAACATTTA
Phenotype
MIM: 605688 MIM: 162080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPNE6 PubMed Links

Gene Details

Gene
CPNE6
Gene Name
copine 6
There are no transcripts associated with this gene.

Gene
NRL
Gene Name
neural retina leucine zipper
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006177.3 1747 UTR 3 NP_006168.1
XM_005267708.4 1747 UTR 3 XP_005267765.1
XM_005267709.3 1747 UTR 3 XP_005267766.1
XM_005267710.3 1747 UTR 3 XP_005267767.1
XM_011536801.2 1747 UTR 3 XP_011535103.2
XM_011536802.1 1747 UTR 3 XP_011535104.1
XM_011536804.2 1747 UTR 3 XP_011535106.1
XM_011536805.2 1747 UTR 3 XP_011535107.1
XM_011536806.2 1747 UTR 3 XP_011535108.2
XM_017021350.1 1747 UTR 3 XP_016876839.1

View Full Product Details