Product Details
- SNP ID
-
rs80289104
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:88572080 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCAACATCTTTGATAGTAACGTG[C/G]CTTCAAACAAGAGCAATTTCAGTCG
- Phenotype
-
MIM: 613279
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZC3H14
PubMed Links
Gene Details
- Gene
- ZC3H14
- Gene Name
- zinc finger CCCH-type containing 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001160103.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001153575.1 |
NM_001160104.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001153576.1 |
NM_001326295.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313224.1 |
NM_001326296.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313225.1 |
NM_001326297.1 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_001313226.1 |
NM_001326298.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313227.1 |
NM_001326299.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313228.1 |
NM_001326300.1 |
506 |
UTR 5 |
|
|
NP_001313229.1 |
NM_001326301.1 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_001313230.1 |
NM_001326302.1 |
506 |
UTR 5 |
|
|
NP_001313231.1 |
NM_001326303.1 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_001313232.1 |
NM_001326304.1 |
506 |
UTR 5 |
|
|
NP_001313233.1 |
NM_001326305.1 |
506 |
UTR 5 |
|
|
NP_001313234.1 |
NM_001326306.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313235.1 |
NM_001326307.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313236.1 |
NM_001326308.1 |
506 |
UTR 5 |
|
|
NP_001313237.1 |
NM_001326309.1 |
506 |
UTR 5 |
|
|
NP_001313238.1 |
NM_001326310.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313239.1 |
NM_001326311.1 |
506 |
UTR 5 |
|
|
NP_001313240.1 |
NM_001326312.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313241.1 |
NM_001326313.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_001313242.1 |
NM_001326314.1 |
506 |
UTR 5 |
|
|
NP_001313243.1 |
NM_001326315.1 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_001313244.1 |
NM_001326316.1 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_001313245.1 |
NM_024824.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_079100.2 |
NM_207660.3 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
NP_997543.1 |
NM_207661.2 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
NP_997544.1 |
NM_207662.3 |
506 |
Intron |
|
|
NP_997545.2 |
XM_005268067.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_005268124.1 |
XM_005268068.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_005268125.1 |
XM_005268069.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_005268126.1 |
XM_005268070.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_005268127.1 |
XM_005268071.4 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_005268128.1 |
XM_005268073.3 |
506 |
Intron |
|
|
XP_005268130.1 |
XM_006720257.2 |
506 |
Intron |
|
|
XP_006720320.1 |
XM_011537160.2 |
506 |
Missense Mutation |
CCT,GCT |
P62A |
XP_011535462.1 |
XM_011537161.2 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_011535463.1 |
XM_011537162.2 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_011535464.1 |
XM_017021648.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_016877137.1 |
XM_017021649.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_016877138.1 |
XM_017021650.1 |
506 |
Missense Mutation |
CCT,GCT |
P96A |
XP_016877139.1 |
View Full Product Details