Product Details

SNP ID
rs80106218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:26874104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCGACTCCCTTCAAGGGGAGAA[A/G]GAGCATGTGGGCTTTCGGGTTTTTA
Phenotype
MIM: 137142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRA5 PubMed Links

Gene Details

Gene
GABRA5
Gene Name
gamma-aminobutyric acid type A receptor alpha5 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000810.3 Intron NP_000801.1
NM_001165037.1 Intron NP_001158509.1
XM_005268258.2 Intron XP_005268315.1
XM_006720459.2 Intron XP_006720522.1
XM_017022055.1 Intron XP_016877544.1
XM_017022056.1 Intron XP_016877545.1

View Full Product Details