Product Details

SNP ID
rs78876080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140185760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAAAGTGTCCATATCAGGAGCA[A/G]GTTCTTCCTAGGAGAAGGCCTACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYSTM1 PubMed Links

Gene Details

Gene
CYSTM1
Gene Name
cysteine rich transmembrane module containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032412.3 Intron NP_115788.1

View Full Product Details