Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032148.3 | 3774 | UTR 3 | NP_115524.3 | ||
XM_005269176.1 | 3774 | UTR 3 | XP_005269233.1 | ||
XM_005269178.1 | 3774 | UTR 3 | XP_005269235.1 | ||
XM_005269179.1 | 3774 | UTR 3 | XP_005269236.1 | ||
XM_006719630.2 | 3774 | UTR 3 | XP_006719693.1 | ||
XM_011538807.2 | 3774 | UTR 3 | XP_011537109.1 | ||
XM_011538808.1 | 3774 | UTR 3 | XP_011537110.1 | ||
XM_011538809.1 | 3774 | UTR 3 | XP_011537111.1 | ||
XM_011538810.2 | 3774 | UTR 3 | XP_011537112.1 | ||
XM_011538811.2 | 3774 | UTR 3 | XP_011537113.1 | ||
XM_011538813.2 | 3774 | UTR 3 | XP_011537115.1 | ||
XM_017020013.1 | 3774 | UTR 3 | XP_016875502.1 | ||
XM_017020014.1 | 3774 | UTR 3 | XP_016875503.1 | ||
XM_017020015.1 | 3774 | Intron | XP_016875504.1 | ||
XM_017020016.1 | 3774 | UTR 3 | XP_016875505.1 | ||
XM_017020017.1 | 3774 | UTR 3 | XP_016875506.1 |