Product Details

SNP ID
rs76696077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:77977052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATTCCAGCATGACCAAGGAATG[A/G]AAAATCCAAGAATGTGCCCTGGCCT
Phenotype
MIM: 614258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR3A PubMed Links

Gene Details

Gene
POLR3A
Gene Name
RNA polymerase III subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007055.3 4732 UTR 3 NP_008986.2

View Full Product Details