Product Details

SNP ID
rs78087466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48437714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCATGCACCTTTGGCTGCTAAT[G/T]GTTCCTGACGTGCACTCTTCCGAGT
Phenotype
MIM: 610585 MIM: 601158
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP22 PubMed Links

Gene Details

Gene
ARHGAP22
Gene Name
Rho GTPase activating protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256024.1 4201 Intron NP_001242953.1
NM_001256025.2 4201 Intron NP_001242954.1
NM_001256026.1 4201 Intron NP_001242955.1
NM_021226.3 4201 Intron NP_067049.2
XM_005270014.3 4201 Intron XP_005270071.1
XM_011540002.1 4201 Intron XP_011538304.1
XM_011540003.1 4201 Intron XP_011538305.1
XM_011540004.1 4201 Intron XP_011538306.1
XM_011540005.1 4201 Intron XP_011538307.1
XM_011540006.1 4201 Intron XP_011538308.1
XM_011540007.2 4201 Intron XP_011538309.1
XM_011540011.2 4201 Intron XP_011538313.1
XM_011540012.2 4201 Intron XP_011538314.1
XM_011540013.2 4201 Intron XP_011538315.1
XM_011540015.2 4201 Intron XP_011538317.1
XM_017016470.1 4201 Intron XP_016871959.1
XM_017016471.1 4201 Intron XP_016871960.1
XM_017016472.1 4201 Intron XP_016871961.1
XM_017016473.1 4201 Intron XP_016871962.1
XM_017016474.1 4201 Intron XP_016871963.1
XM_017016475.1 4201 Intron XP_016871964.1
XM_017016476.1 4201 Intron XP_016871965.1
XM_017016477.1 4201 Intron XP_016871966.1
Gene
MAPK8
Gene Name
mitogen-activated protein kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278547.1 4201 UTR 3 NP_001265476.1
NM_001278548.1 4201 UTR 3 NP_001265477.1
NM_001323302.1 4201 UTR 3 NP_001310231.1
NM_001323320.1 4201 UTR 3 NP_001310249.1
NM_001323321.1 4201 UTR 3 NP_001310250.1
NM_001323322.1 4201 UTR 3 NP_001310251.1
NM_001323323.1 4201 UTR 3 NP_001310252.1
NM_001323324.1 4201 UTR 3 NP_001310253.1
NM_001323325.1 4201 UTR 3 NP_001310254.1
NM_001323326.1 4201 UTR 3 NP_001310255.1
NM_001323327.1 4201 UTR 3 NP_001310256.1
NM_001323328.1 4201 UTR 3 NP_001310257.1
NM_001323329.1 4201 UTR 3 NP_001310258.1
NM_001323330.1 4201 UTR 3 NP_001310259.1
NM_001323331.1 4201 UTR 3 NP_001310260.1
NM_139046.3 4201 UTR 3 NP_620634.1
NM_139049.3 4201 UTR 3 NP_620637.1

View Full Product Details