Product Details
- SNP ID
-
rs75071437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97738631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACAAGAGGCTGGAGATCTACCTG[A/G]AACCCTTGAAGGATGCAGGTGATGG
- Phenotype
-
MIM: 610243
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFYVE27
PubMed Links
Gene Details
- Gene
- ZFYVE27
- Gene Name
- zinc finger FYVE-type containing 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001002261.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_001002261.1 |
NM_001002262.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_001002262.1 |
NM_001174119.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_001167590.1 |
NM_001174120.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_001167591.1 |
NM_001174121.1 |
356 |
Intron |
|
|
NP_001167592.1 |
NM_001174122.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_001167593.1 |
NM_144588.6 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
NP_653189.3 |
XM_005269502.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269559.1 |
XM_005269503.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269560.1 |
XM_005269504.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269561.1 |
XM_005269505.4 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269562.1 |
XM_005269506.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269563.1 |
XM_005269508.3 |
356 |
UTR 5 |
|
|
XP_005269565.1 |
XM_005269509.3 |
356 |
UTR 5 |
|
|
XP_005269566.1 |
XM_005269510.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269567.1 |
XM_005269511.3 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_005269568.1 |
XM_011539252.2 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_011537554.1 |
XM_011539253.2 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_011537555.1 |
XM_011539254.2 |
356 |
UTR 5 |
|
|
XP_011537556.1 |
XM_011539255.2 |
356 |
UTR 5 |
|
|
XP_011537557.1 |
XM_017015644.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871133.1 |
XM_017015645.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871134.1 |
XM_017015646.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871135.1 |
XM_017015647.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871136.1 |
XM_017015648.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871137.1 |
XM_017015649.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871138.1 |
XM_017015650.1 |
356 |
UTR 5 |
|
|
XP_016871139.1 |
XM_017015651.1 |
356 |
UTR 5 |
|
|
XP_016871140.1 |
XM_017015652.1 |
356 |
UTR 5 |
|
|
XP_016871141.1 |
XM_017015653.1 |
356 |
UTR 5 |
|
|
XP_016871142.1 |
XM_017015654.1 |
356 |
UTR 5 |
|
|
XP_016871143.1 |
XM_017015655.1 |
356 |
Missense Mutation |
AAA,GAA |
K52E |
XP_016871144.1 |
View Full Product Details