Product Details
- SNP ID
-
rs77733832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:101090948 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGGGGAAGTGCCTGGGGTGTGG[A/T]GAGGGTGTGGGCTTCAGGGAAGAGG
- Phenotype
-
MIM: 612734
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TLX1NB
PubMed Links
Gene Details
- Gene
- TLX1NB
- Gene Name
- TLX1 neighbor
There are no transcripts associated with this gene.
View Full Product Details