Product Details

SNP ID
rs78061258
Assay Type
Functionally Tested
NCBI dbSNP Submissions
20
Location
Chr.1:42177864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATTACTCAACCATAGAATCACT[A/G]AGGCTAATTAAAGTGATTTAGCATG
Phenotype
MIM: 616035
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FOXJ3 PubMed Links

Gene Details

Gene
FOXJ3
Gene Name
forkhead box J3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198850.1 3931 UTR 3 NP_001185779.1
NM_001198851.1 3931 UTR 3 NP_001185780.1
NM_001198852.1 3931 UTR 3 NP_001185781.1
NM_014947.4 3931 UTR 3 NP_055762.3
XM_005270632.4 3931 UTR 3 XP_005270689.1
XM_006710458.3 3931 UTR 3 XP_006710521.1
XM_006710459.3 3931 UTR 3 XP_006710522.1
XM_011541026.2 3931 UTR 3 XP_011539328.1
XM_017000693.1 3931 UTR 3 XP_016856182.1
XM_017000694.1 3931 UTR 3 XP_016856183.1
XM_017000695.1 3931 UTR 3 XP_016856184.1

View Full Product Details