Product Details

SNP ID
rs78064220
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:43302101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTGGAGGGCCATTTTCAGGGG[G/T]TGTTCATAGGGTAATTGTGTTGGAT
Phenotype
MIM: 600222
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIE1 PubMed Links

Gene Details

Gene
TIE1
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253357.1 Intron NP_001240286.1
NM_005424.4 Intron NP_005415.1
XM_005271163.1 Intron XP_005271220.1
XM_006710869.1 Intron XP_006710932.1
XM_017002207.1 Intron XP_016857696.1

View Full Product Details