Product Details

SNP ID
rs75547379
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:43451633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTGAAAGGGTGGGAGGGCAAA[C/G]GAAGGTCCTCTCACCAACAATGGGC
Phenotype
MIM: 615463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HYI PubMed Links

Gene Details

Gene
HYI
Gene Name
hydroxypyruvate isomerase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190880.2 11365 Intron NP_001177809.1
NM_001243526.1 11365 Intron NP_001230455.1
NM_031207.5 11365 Intron NP_112484.3
XM_005271239.4 11365 Intron XP_005271296.1
XM_005271240.4 11365 Intron XP_005271297.1
XM_006710937.3 11365 Intron XP_006711000.1
XM_011542222.2 11365 Intron XP_011540524.1
Gene
MIR6735
Gene Name
microRNA 6735
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 11365 UTR 3 NP_056099.3
XM_005270686.3 11365 Intron XP_005270743.1
XM_006710501.3 11365 Intron XP_006710564.1
XM_011541106.2 11365 Intron XP_011539408.1
XM_011541107.2 11365 Intron XP_011539409.1
XM_017000819.1 11365 Intron XP_016856308.1
XM_017000820.1 11365 Intron XP_016856309.1
XM_017000821.1 11365 Intron XP_016856310.1

View Full Product Details