Product Details

SNP ID
rs77848815
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:32222386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGACCCATAATCCGGAAGTGAC[C/T]TCGAAACCTTTTCCGGTCTTACTCA
Phenotype
MIM: 603911
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCDC2B PubMed Links

Gene Details

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003757.3 478 UTR 5 NP_003748.1
XM_017002671.1 478 UTR 5 XP_016858160.1
Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 478 Intron NP_061991.3
XM_011541795.2 478 Intron XP_011540097.1
XM_017001812.1 478 Intron XP_016857301.1
XM_017001813.1 478 Intron XP_016857302.1
XM_017001814.1 478 Intron XP_016857303.1
XM_017001815.1 478 Intron XP_016857304.1
XM_017001816.1 478 Intron XP_016857305.1
XM_017001817.1 478 Intron XP_016857306.1
XM_017001818.1 478 Intron XP_016857307.1
XM_017001819.1 478 Intron XP_016857308.1
XM_017001820.1 478 Intron XP_016857309.1
XM_017001821.1 478 Intron XP_016857310.1
XM_017001822.1 478 Intron XP_016857311.1
XM_017001823.1 478 Intron XP_016857312.1
XM_017001824.1 478 Intron XP_016857313.1
XM_017001825.1 478 Intron XP_016857314.1
XM_017001826.1 478 Intron XP_016857315.1
XM_017001827.1 478 Intron XP_016857316.1
XM_017001828.1 478 Intron XP_016857317.1
XM_017001829.1 478 Intron XP_016857318.1
XM_017001830.1 478 Intron XP_016857319.1
XM_017001831.1 478 Intron XP_016857320.1

View Full Product Details