Product Details

SNP ID
rs79147717
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:107148162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTCAGATACTGATATCTCAGCC[C/T]GCTTGAGCATCCCTTGTGAGCTGTG
Phenotype
MIM: 608818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTNG1 PubMed Links

Gene Details

Gene
NTNG1
Gene Name
netrin G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113226.2 1329 UTR 5 NP_001106697.1
NM_001113228.2 1329 UTR 5 NP_001106699.1
NM_001312688.1 1329 UTR 5 NP_001299617.1
NM_014917.3 1329 UTR 5 NP_055732.2
XM_006710455.2 1329 UTR 5 XP_006710518.1
XM_006710456.3 1329 UTR 5 XP_006710519.1
XM_011541021.1 1329 UTR 5 XP_011539323.1
XM_011541024.2 1329 UTR 5 XP_011539326.1
XM_011541025.1 1329 UTR 5 XP_011539327.1
XM_017000680.1 1329 UTR 5 XP_016856169.1
XM_017000681.1 1329 UTR 5 XP_016856170.1
XM_017000682.1 1329 UTR 5 XP_016856171.1
XM_017000683.1 1329 UTR 5 XP_016856172.1
XM_017000684.1 1329 UTR 5 XP_016856173.1
XM_017000685.1 1329 UTR 5 XP_016856174.1
XM_017000686.1 1329 UTR 5 XP_016856175.1

View Full Product Details