Product Details
- SNP ID
-
rs75051239
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:110403232 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCTTTTTACAAGGCTGTGATAG[C/T]AGCAGTTGTATATATATACTTTTTT
- Phenotype
-
MIM: 608521
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAMTOR5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57125530] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAMTOR5
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006402.2 |
|
Intron |
|
|
NP_006393.2 |
- Gene
- LAMTOR5-AS1
- Gene Name
- LAMTOR5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details