Product Details

SNP ID
rs76179044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125958845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTAAATCAAAATGGCTGTCTA[C/T]GGACAATGACTCCCCAGTTTTCTAA
Phenotype
MIM: 608707
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDON PubMed Links

Gene Details

Gene
CDON
Gene Name
cell adhesion associated, oncogene regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243597.1 6050 UTR 3 NP_001230526.1
NM_016952.4 6050 UTR 3 NP_058648.4
XM_011542862.2 6050 UTR 3 XP_011541164.1
XM_011542863.2 6050 UTR 3 XP_011541165.1
XM_011542864.2 6050 UTR 3 XP_011541166.1
XM_011542865.2 6050 UTR 3 XP_011541167.1
XM_011542866.2 6050 UTR 3 XP_011541168.1
XM_017017873.1 6050 UTR 3 XP_016873362.1
Gene
LOC338667
Gene Name
V-set and immunoglobulin domain-containing protein 10-like
There are no transcripts associated with this gene.

View Full Product Details