Product Details

SNP ID
rs76175248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134249157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCTGTCATGAAATAGAAAGCA[C/T]AGTCTTTACCATGGACCACTTAGGG
Phenotype
MIM: 604773 MIM: 613739 MIM: 610027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAD8 PubMed Links

Gene Details

Gene
ACAD8
Gene Name
acyl-CoA dehydrogenase family member 8
There are no transcripts associated with this gene.

Gene
THYN1
Gene Name
thymocyte nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037304.1 Intron NP_001032381.1
NM_001037305.1 Intron NP_001032382.1
NM_014174.2 Intron NP_054893.1
NM_199297.1 Intron NP_954994.1
NM_199298.1 Intron NP_954995.1
XM_005271527.3 Intron XP_005271584.1
XM_005271528.3 Intron XP_005271585.1
Gene
VPS26B
Gene Name
VPS26, retromer complex component B
There are no transcripts associated with this gene.

View Full Product Details