Product Details

SNP ID
rs77825133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:94705888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGTCATTTTAAACATGATACA[C/T]TAAATAAAGCATGTTGAGGTCAAAG
Phenotype
MIM: 616296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCTP1 PubMed Links

Gene Details

Gene
MCTP1
Gene Name
multiple C2 and transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002796.3 4009 UTR 3 NP_001002796.1
NM_001297777.1 4009 UTR 3 NP_001284706.1
NM_024717.5 4009 UTR 3 NP_078993.4
XM_005272082.4 4009 UTR 3 XP_005272139.1
XM_005272090.4 4009 UTR 3 XP_005272147.1
XM_006714692.3 4009 UTR 3 XP_006714755.1
XM_011543649.2 4009 UTR 3 XP_011541951.1
XM_011543650.2 4009 UTR 3 XP_011541952.1
XM_017009855.1 4009 UTR 3 XP_016865344.1
XM_017009856.1 4009 Intron XP_016865345.1
XM_017009857.1 4009 UTR 3 XP_016865346.1
XM_017009858.1 4009 UTR 3 XP_016865347.1
XM_017009859.1 4009 UTR 3 XP_016865348.1
XM_017009860.1 4009 UTR 3 XP_016865349.1
XM_017009861.1 4009 UTR 3 XP_016865350.1
XM_017009862.1 4009 UTR 3 XP_016865351.1
XM_017009863.1 4009 UTR 3 XP_016865352.1
XM_017009864.1 4009 UTR 3 XP_016865353.1
XM_017009865.1 4009 Intron XP_016865354.1
Gene
SLF1
Gene Name
SMC5-SMC6 complex localization factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032290.3 4009 Intron NP_115666.2

View Full Product Details