Product Details

SNP ID
rs79629586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132376475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGATGTCTGCTGAACGCAAAGA[C/T]GCATTTCATCACAGCCCAGGGTACG
Phenotype
MIM: 600777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTF1 PubMed Links

Gene Details

Gene
TTF1
Gene Name
transcription termination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205296.1 Intron NP_001192225.1
NM_007344.3 Intron NP_031370.2
XM_006717273.3 Intron XP_006717336.2
XM_017015105.1 Intron XP_016870594.1

View Full Product Details