Product Details

SNP ID
rs75043108
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:211386881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCAGATGACAATTATATAGTA[A/T]GTCTCAAGAAGTCTATTTAAAAAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details